Learn More
Medchemexpress LLC Methyl 6-O-(N-heptylcarbamoyl)-α-d-glucopyranoside | 115457-83-5 | MFCD00077425 | ≥97.0% | 335.39 g·mol⁻¹ | C15H29NO7 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW2501825G
Encompass_Preferred
Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside (Hecameg) is a naturally derived biochemical reagent used as a precursor or raw material for plant growth regulators and agricultural formulations, and as a reagent or biomaterial in life-science research. It is supplied as a white to off-white powder; molecular formula C15H29NO7; molecular weight 335.39 g·mol⁻¹.
  • Used as a precursor for plant growth regulators and fertilizer components.
  • Enhances nutrient absorption and promotes plant development in formulations.
  • Suitable as a biochemical reagent or biomaterial for research applications.
  • White to off-white powder appearance for easy handling and formulation.
  • Available in laboratory pack sizes for experimental use.
Catalog No. 50-002-80882