Learn More
Medchemexpress LLC Methyl 6-O-(N-heptylcarbamoyl)-α-d-glucopyranoside | 115457-83-5 | MFCD00077425 | >99.0% | 335.39 g·mol⁻1 | C15H29NO7 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW2501821G
Encompass_Preferred
Methyl 6-O-(N-heptylcarbamoyl)-α-d-glucopyranoside is an alkyl glycoside surfactant (commonly called Hecameg) used as a raw material in plant growth regulators and agricultural formulations. It acts as a non-ionic surfactant to improve nutrient absorption and formulation performance, and is also used in laboratory research for gentle solubilization of membrane-associated biomolecules.
  • Used as raw material for plant growth regulators and fertilizers.
  • Non-ionic surfactant that enhances nutrient absorption in plants.
  • Improves formulation wetting and spreading properties.
  • Suitable for gentle solubilization of membrane-bound proteins in research.
  • Available in small-scale laboratory packaging for formulation testing.
Catalog No. 50-002-80529