Learn More
Medchemexpress LLC Methyl 9(Z),11(E)-octadecadienoate | 13058-52-1 | 99.2% | 294.47 g/mol | C19H34O2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12437025MG
Encompass_Preferred
9(Z),11(E)-Conjugated linoleic acid methyl ester (methyl 9(Z),11(E)-octadecadienoate) is a high-purity methyl ester isomer of linoleic acid used as a research reference standard for analytical and biochemical studies. Supplied as a colorless liquid, it is suitable for chromatographic and spectroscopic analysis, lipid profiling, and method development. Store protected from light under inert gas at low temperatures to maintain stability.
  • High purity (99.2%) suitable for analytical applications.
  • Available in small package sizes for standards and assays.
  • Liquid form compatible with chromatographic and spectroscopic methods.
  • Stable when stored under nitrogen and protected from light at low temperatures.
  • Applicable as an isomeric reference for conjugated linoleic acid analysis.
Catalog No. 50-002-36874