Learn More
Medchemexpress LLC Methyl 9H-xanthene-9-carboxylate | 39497-06-8 | 95.0% | C15H12O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY78996100MG
Encompass_Preferred
Methyl 9H-xanthene-9-carboxylate is a chemical compound used as a drug intermediate for the synthesis of various active compounds. It is intended for laboratory use only and not for patient administration.
  • Used as a drug intermediate
  • Suitable for synthesis of various active compounds
  • Intended for laboratory chemicals and manufacturing
  • For research use only
Catalog No. 50-004-35097