Learn More
U.S. Pharmacopeia Methyl Alcohol, 67-56-1, MFCD00004595, 3 x 1.5 mL
Supplier:  U.S. Pharmacopeia 1424109
Encompass
Molecular formula CH4O, Molecular weight 32.05, Melting Point -144.04 °F (-97.8 °C), Boiling Point 148.46 °F (64.7 °C), Synonyms: Wood alcohol, Methanol
Catalog No. NC9681149