Learn More
Medchemexpress LLC Methyl aminolevulinate hydrochloride | 79416-27-6 | 99.0% | C6H12ClNO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA0169A10MM/1ML
Encompass_Preferred
Methyl aminolevulinate hydrochloride is a chemical compound primarily recognized as a sensitizer in photodynamic therapy (PDT). It functions as a proagent, metabolizing to Protoporphyrin IX, and is valuable for various research applications in disease areas such as cancer and inflammatory conditions.
  • Acts as a sensitizer in photodynamic therapy (PDT)
  • Metabolized into Protoporphyrin IX
  • Relevant for cancer research
  • Explored in inflammation and immune system disease studies
  • Associated with Reactive Oxygen Species (ROS) pathways
  • Applied in studies for acne vulgaris and basal cell carcinoma
Catalog No. 50-004-32700