Learn More
Selleck Chemical LLC Methyl-B-cyclodextrin 100mg
SDP
Supplier:  Selleck Chemical LLC S6827100MG
Encompass_Preferred
Methyl-?-cyclodextrin (M?CD, Methyl-beta-cyclodextrin, M-?-CyD, Methyl-?-CD) is acholesterol-removing agent mainly used forlipid raft disruption. Methyl-?-cyclodextrin induces caspase-dependent apoptosis in PEL cells and inhibits the growth and invasion of PEL cells without apparent adverse effects, suggesting the potential effective antitumor activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3225