Learn More
Medchemexpress LLC Methyl benzyl-L-serinate | 123639-56-5 | 209.25 | 1 G

Supplier: Medchemexpress LLC HYW003903A1G
Methyl benzyl-L-serinate is a serine derivative that appears as an off-white to light yellow solid-liquid mixture. It is intended for research use only.
- High purity: 98.19%
- Off-white to light yellow solid-liquid mixture
- Soluble in DMSO at concentrations of ≥ 100 mg/mL
- Shipped at room temperature
- Storage at 4°C protected from light (in solid form)
- In solvent, store at -80°C for up to 6 months or -20°C for 1 month protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.