Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl benzyl-L-serinate | 123639-56-5 | 209.25 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW003903A10MM/1ML

Encompass_Preferred

Methyl benzyl-L-serinate is a serine derivative. It is an off-white to light yellow solid-liquid mixture, intended for research use only.

  • Serine derivative
  • Off-white to light yellow solid-liquid mixture
  • Purity of 98.19%
  • Molecular weight of 209.25
  • Intended for research use only

Catalog No. 50-004-49111


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.