Learn More
Medchemexpress LLC Methyl benzyl-L-serinate | 123639-56-5 | 209.25 | 100 MG

Supplier: Medchemexpress LLC HYW003903A100MG
Methyl benzyl-L-serinate is a serine derivative. It is supplied as a solid-liquid mixture, with an off-white to light yellow color.
- Has a molecular weight of 209.25
- Purity of 98.19%
- Ships at room temperature
- Store at 4°C, protected from light
- Soluble in DMSO (≥ 100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.