Learn More
Medchemexpress LLC Methyl benzyl-L-serinate | 123639-56-5 | 209.25 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW003903A
Methyl benzyl-L-serinate is a serine derivative intended for research use only. This product is provided as an off-white to light yellow solid-liquid mixture.
- Appearance: Off-white to light yellow solid-liquid mixture
- Storage: 4°C, protect from light
- In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)
- Solubility in DMSO: ≥ 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.