Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl benzyl-L-serinate | 123639-56-5 | 209.25 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW003903A5G

Encompass_Preferred

Methyl benzyl-L-serinate is a derivative of serine, designed exclusively for research applications.

  • Formula: C11H15NO3
  • Appearance: Solid-liquid mixture
  • Color: Off-white to light yellow
  • Shipping: Room temperature within the continental US; may vary elsewhere
  • Storage: 4°C, protected from light. In solvent: -80°C for 6 months; -20°C for 1 month (protected from light)

Catalog No. 50-004-49244


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.