Learn More
Medchemexpress LLC Methyl ((benzyloxy)carbonyl)-D-serinate | 93204-36-5 | 253.25 g·mol⁻¹ | C12H15NO5 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0523101KG
Encompass_Preferred
Methyl ((benzyloxy)carbonyl)-D-serinate is a Cbz-protected D-serine methyl ester used as a building block in peptide synthesis and amino-acid derivative chemistry. It is supplied as a white to off-white solid and is intended for research use, with storage recommendations for powder and solutions provided by standard laboratory practice.
  • Cbz-protected D-serine methyl ester suitable for peptide synthesis and derivatization.
  • White to off-white solid appearance.
  • Available in multiple pack sizes, including bulk quantities up to 1 kg.
  • Documented storage stability for powder and solutions to support long-term use.
  • Includes solubility and documentation sections for experimental planning.
Catalog No. 50-002-10016