Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl ((benzyloxy)carbonyl)-D-serinate | 93204-36-5 | 253.25 g/mol | C12H15NO5 | 50 G
SDP

Supplier:  Medchemexpress LLC HYW05231050G

Encompass_Preferred

Methyl ((benzyloxy)carbonyl)-D-serinate is a protected D-serine derivative used as a building block in organic synthesis and research applications. It is supplied as a white to off-white solid and intended for research use only.

  • Protected D-serine derivative for synthetic and research applications.
  • Molecular formula: C12H15NO5.
  • Molecular weight: 253.25 g/mol.
  • CAS number: 93204-36-5.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years).
  • Available package sizes: 5 g-1 kg.

Catalog No. 50-002-09982


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.