Learn More
Medchemexpress LLC Methyl ((benzyloxy)carbonyl)-D-serinate | 93204-36-5 | MFCD00799501 | 253.25 g/mol | C12H15NO5 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW052310100G
Encompass_Preferred
Methyl ((benzyloxy)carbonyl)-D-serinate is a Cbz-protected D-serine methyl ester used as a synthetic intermediate and building block in peptide and organic synthesis.
  • Cbz-protected amino acid methyl ester suitable for synthetic transformations.
  • Serves as a building block in peptide synthesis and small-molecule assembly.
  • Typically supplied as a white to off-white solid for ease of handling.
  • Available in multiple pack sizes to support small-scale and bulk use.
Catalog No. 50-002-10921