Learn More
Medchemexpress LLC Methyl ((benzyloxy)carbonyl)-D-serinate | 93204-36-5 | MFCD00799501 | >98.0% | 253.25 g·mol⁻1 | C12H15NO5 | 25 G

Supplier: Medchemexpress LLC 25GHYW052310
Methyl ((benzyloxy)carbonyl)-D-serinate is a Cbz-protected D-serine methyl ester supplied as a white to off-white solid for laboratory research use only. It is used as a protected serine building block and intermediate in organic synthesis, peptide chemistry, and medicinal chemistry applications.
- Appearance: white to off-white solid.
- Molecular weight approximately 253.25 g·mol⁻1.
- Chemical formula: C12H15NO5.
- CAS number: 93204-36-5.
- Typical reported purity: >98.0% (verify with COA).
- Common package sizes: 5 g, 10 g, 25 g, 50 g, 100 g, 500 g, 1 kg.
- Storage: powder -20°C for long term; in solvent store at -80°C for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.