Learn More
Medchemexpress LLC Methyl ((benzyloxy)carbonyl)-D-serinate | 93204-36-5 | MFCD00799501 | 99.0% | 253.25 g/mol | C12H15NO5 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW052310500G
Encompass_Preferred
Methyl ((benzyloxy)carbonyl)-D-serinate is a Cbz-protected D-serine methyl ester used as an amino acid derivative and intermediate in peptide and small-molecule synthesis. It is supplied as a solid powder and commonly used where temporary protection of the serine amine is required.
  • Used as an amino acid derivative and peptide synthesis intermediate.
  • Cbz-protected amine provides temporary protection during multi-step synthesis.
  • Supplied as a solid powder for storage and handling convenience.
  • Available in multiple pack sizes, including 500 g.
  • Storage: powder - stable at -20°C for long term and 4°C for shorter term.
Catalog No. 50-002-10940