Learn More
Medchemexpress LLC Methyl-beta-cyclodextrin | 128446-36-6 | MFCD00074980 | 100.0% | 1310 g/mol (average) | C54H94O35 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1014615G
Encompass_Preferred
Methyl-β-cyclodextrin is a methylated derivative of β-cyclodextrin supplied as a white to off-white powder for laboratory research. It is commonly used to increase solubility of hydrophobic molecules and to deplete cholesterol from cell membranes in biological studies. The material dissolves in water and DMSO and is offered in multiple lab-scale package sizes.
  • Solubilizes hydrophobic compounds for formulation and assays.
  • Extracts membrane cholesterol for cell biology studies.
  • Water and DMSO soluble at research concentrations.
  • High purity suitable for analytical and biological applications.
  • Available in multiple small lab packaging sizes for flexibility.
Catalog No. 50-002-49173