Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl brassylate | 1472-87-3 | 98.0% | 272.39 | 50 G
SDP

Supplier:  Medchemexpress LLC HYW01163750G

Encompass_Preferred

Methyl brassylate (Dimethyl tridecanedioate) is an ester product intended for research use only and not for sale to patients.

  • Appearance: solid
  • Color: white to off-white
  • Storage: store at room temperature for 3 years; in solvent, store at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-004-55005


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.