Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | 500G
SDP
Supplier:  Medchemexpress LLC HY125861C500G
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether powder used as a thickener, stabilizer, emulsifier, and thermogelating agent. The 1500 mPa·s viscosity grade delivers moderate thickening and heat-induced gel formation, making it suitable for formulation development, laboratory assays, and product prototyping across pharmaceutical, cosmetic, food, and construction applications.
  • Provides moderate viscosity and thermal gelation
  • Water soluble and cold-water dispersible
  • Functions as a thickener, stabilizer, and emulsifier
  • Suitable for pharmaceutical, cosmetic, and food formulations
  • Available as a bulk powder for laboratory and pilot-scale use
Catalog No. 50-002-35722