Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | 98.0% | C6H7O2(OR1)(OR2)(OR3) (polymeric repeat unit) | 25G
SDP
Supplier:  Medchemexpress LLC HY125861D25G
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether supplied as a powder with a nominal viscosity of 40,000 mPa·s. It dissolves in water with warming and ultrasonic treatment to form viscous solutions and gels used as thickeners, stabilizers, and gel-forming agents for laboratory formulations, assay reagents, and viscosity modification. Recommended storage is sealed at 4°C; solutions have specified short-term stability at -20°C and long-term stability at -80°C per datasheet.
  • High-viscosity aqueous thickener for formulation and assay work.
  • Soluble in water (approx. 10 mg/mL) with heating and sonication.
  • Forms thermogel on heating and stabilizes emulsions and suspensions.
  • Supplied as a 25 g powder for laboratory use.
  • Polymeric material with variable molecular weight; no single fixed formula.
  • SDS and technical datasheet available for handling, solubility, and storage details.
Catalog No. 50-002-36114