Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | >95.0% | 454.51 g/mol | C20H38O11 | 25G
SDP
Supplier:  Medchemexpress LLC HY125861A25G
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether supplied as a low-viscosity reagent (15 mPa·s) used to modify rheology and stabilize formulations. It is water soluble, thermogelating, and commonly applied as a thickener, stabilizer, emulsifier, and excipient in pharmaceutical, food, cosmetic, and construction applications. Typical small-pack sizes support laboratory and formulation development work.
  • Low viscosity grade (15 mPa·s) suitable for easy dispersion
  • Water soluble and forms a gel on heating
  • Functions as a thickener, stabilizer, and emulsifier
  • Useful as an excipient in drug delivery and formulation testing
  • Available in small pack sizes for research and development
Catalog No. 50-002-36566