Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | Not available | [OC5H5(OR)2CH2OR]n, R = H or CH3 | 100G
SDP
Supplier:  Medchemexpress LLC HY125861B100G
Encompass_Preferred
Methyl cellulose (viscosity 400 mPa·s) is a non-ionic cellulose ether supplied as a powdered solid used to increase viscosity, stabilize dispersions, and provide thermogelation in aqueous systems. It dissolves in cold water to form viscous solutions and reversibly gels on heating, making it useful for formulation testing and laboratory research.
  • Provides viscosity control in aqueous systems.
  • Acts as a stabilizer and thickening agent.
  • Exhibits thermogelation; gels on heating and liquefies on cooling.
  • Soluble in cold water to form viscous solutions.
  • Off-white to light yellow solid for easy handling.
  • Suitable for research and formulation development applications.
Catalog No. 50-002-35594