Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl cellulose meets USP500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M0430500G
Encompass
Methyl cellulose is an odorless and tasteless material. It is tough and non-toxic. Due to these properties methyl cellulose is used as an adhesive in the food packaging industry. In water-soluble adhesives it is utilized as a thickening agent.
Catalog No. 50-290-2432