Learn More
Medchemexpress LLC Methyl cinnamate | 103-26-4 | MFCD00008458 | 99.6% | 162.19 g/mol | C10H10O2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01721250G
Encompass_Preferred
Methyl cinnamate (methyl 3-phenylpropenoate) is a naturally occurring ester used as a flavoring agent and research reagent. It is colorless to off-white and may be solid or liquid depending on temperature. Molecular formula C10H10O2, molecular weight 162.19 g/mol, and reported purity ~99.6%.
  • Widely used natural flavor compound for sensory studies.
  • Exhibits antimicrobial and tyrosinase-inhibiting activity useful in biochemical research.
  • Suitable for laboratory assays and mechanistic studies involving adipogenesis pathways.
  • High purity supports analytical and preparative applications.
  • Available in common laboratory pack sizes for research use.
Catalog No. 50-003-00569