Learn More
Medchemexpress LLC Methyl α-D-mannopyranoside | 617-04-9 | MFCD00063262 | 98.0% | 194.18 g/mol | C7H14O6 | 5 G

Supplier: Medchemexpress LLC HYW0398975G
Methyl α-D-mannopyranoside is the alpha anomer of methyl D-mannoside used as an analytical standard and research reagent in glycobiology, carbohydrate chemistry, and method development. It is used as a probe in macrophage-targeting studies and for analytical calibration. Supplied for research use only.
- Analytical standard for glycobiology and carbohydrate research.
- Reported use as a probe for macrophage targeting in inhalation studies.
- Purity 98.0% (HPLC).
- Molecular formula C7H14O6; molecular weight 194.18 g/mol.
- Available as a 5 g laboratory pack (typical).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.