Learn More
Medchemexpress LLC Methyl gallate | 99-24-1 | MFCD00002194 | 184.15 | 50 G

Supplier: Medchemexpress LLC HYN201050G
Methyl gallate is a plant phenolic with antioxidant, anticancer, and anti-inflammatory activities. It also exhibits bacterial inhibition activity and has anti-HIV-1 and HIV-1 enzyme inhibitory activities.
- Purity: 99.97%
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants, Paeoniaceae, Paeonia lactiflora Pall.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.