Learn More
Electron Microscopy Sciences Methyl Green, Certified, C.I. DcG-10 25 GR

Supplier: Electron Microscopy Sciences 18700
(Zinc Chloride Salt; Ethyl Green)
CAS #7114-03-6 C₂₇H₃₅N₃C₁₂·ZnC₁₂ F.W. 608.79
Solubility: 8.0% water; 3.0% Alcohol; 1.8% Cellosolve; 3.5% Glycol; 0.0% Xylene
Used as a counterstain in Almann's aniline acid fuchsin technique for mitochrondria; used in Kurnick's Methyl Green-Pyonin stain for nuclei and cystoplasmic granules of liver cells. Marker for DNA at acid and neutral conditions (J.B. Boyd, H.R. Mitchell, 1965 Anal. Biochem., 13, 28). Used for epoxy-embedded thick sections (Sievers, J. 1971). Basic two-dye stains for epoxy-embedded 0.3-1 micron sections (Stain Technol., 46:195)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.