Learn More
Medchemexpress LLC Methyl homoveratrate | 15964-79-1 | MFCD00017206 | 98.3% | 210.23 g/mol | C11H14O4 | 10 G

Supplier: Medchemexpress LLC HYW04203910G
Methyl homoveratrate is a research chemical and an in vivo metabolite of RWJ-26240, used as a reference compound in studies of voltage-sensitive calcium channels. The substance is supplied as a solid-liquid mixture and is accompanied by COA and SDS documentation.
- Purity: 98.3%.
- Cas number: 15964-79-1.
- Molecular formula: C11H14O4.
- Molecular weight: 210.23 g/mol.
- Appearance: solid-liquid mixture, white to light yellow.
- Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Recommended solvent/stock: DMSO; example stock 10 mM in DMSO.
- Available pack sizes: 1 g, 5 g, 10 g, 25 g, 50 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.