Learn More
Medchemexpress LLC Methyl indole-3-carboxylate | 942-24-5 | 99.9% | C10H9NO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7963510MM/1ML
Encompass_Preferred
Methyl indole-3-carboxylate is a Sorangium metabolite that exhibits weak activity against Gram-positive Nocardia sp. with a minimum inhibitory concentration (MIC) of 33.3 μg/mL. It does not show significant cytotoxicity, with IC50 values of >56.8 μM against L929, PC-3, and KB3-1 cells.
  • Sorangium metabolite
  • Weak activity against Gram-positive Nocardia sp.
  • Does not show significant cytotoxicity against L929, PC-3, and KB3-1 cells (IC50 values > 56.8 μM)
  • Solid appearance
  • Color is off-white to pink
  • For research use only
Catalog No. 50-004-33031