Learn More
Medchemexpress LLC Methyl indole-3-carboxylate | 942-24-5 | 99.9% | C10H9NO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HY79635100G
Encompass_Preferred
Methyl indole-3-carboxylate is a Sorangium metabolite that shows weak activity against Gram-positive Nocardia sp. The compound does not exhibit significant cytotoxicity against various cell lines, making it suitable for certain research applications.
  • Sorangium metabolite
  • Weak activity against Gram-positive Nocardia sp.
  • Minimal cytotoxicity against L929, PC-3, and KB3-1 cells
Catalog No. 50-004-33262