Learn More
Medchemexpress LLC Methyl indole-3-carboxylate | 942-24-5 | 99.9% | C10H9NO2 | 250 G

Supplier: Medchemexpress LLC 250GHY79635
Methyl indole-3-carboxylate is a Sorangium metabolite demonstrating weak activity against Gram-positive Nocardia sp. with a MIC of 33.3 μg/mL. It exhibits no significant cytotoxicity, with IC50 values of >56.8 μM against L929, PC-3, and KB3-1 cells. This compound is supplied as an off-white to pink solid.
- Weak activity against Gram-positive Nocardia sp.
- Not significantly cytotoxic
- Molecular weight of 175.19
- Appearance as an off-white to pink solid
- Store as powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.