Learn More
Medchemexpress LLC Methyl indole-3-carboxylate | 942-24-5 | MFCD00189407 | 99.87% | C10H9NO2 | 500 G
SDP
Supplier:  Medchemexpress LLC HY79635500G
Encompass_Preferred
Methyl indole-3-carboxylate is identified as a Sorangium metabolite. It demonstrates weak activity against Gram-positive Nocardia sp. with a Minimum Inhibitory Concentration (MIC) of 33.3 μg/mL. The compound shows minimal cytotoxicity, with IC50 values exceeding 56.8 μM against L929, PC-3, and KB3-1 cells. This chemical is suitable for various research and analytical applications.
  • Sorangium metabolite
  • Exhibits weak activity against Gram-positive Nocardia sp.
  • Low cytotoxicity against L929, PC-3, and KB3-1 cells
  • Suitable for research and analytical applications
Catalog No. 50-004-33261