Learn More
Medchemexpress LLC Methyl linolenate | 301-00-8 | 99.46% | 292.46 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY21268500MG
Encompass_Preferred
Methyl linolenate is identified as a polyunsaturated fatty acid (PUFA). It exhibits anti-melanogenesis activity with an IC50 of 60 μM. This compound is also valuable for research into the mechanisms and prevention of oxidation and peroxidation of unsaturated fatty acids.
  • Functions as a polyunsaturated fatty acid (PUFA).
  • Demonstrates anti-melanogenesis activity.
  • Useful for studying oxidation/peroxidation mechanisms of unsaturated fatty acids.
  • Down-regulates microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins in B16F10 cells at specific concentrations.
Catalog No. 50-004-03259