Learn More
Medchemexpress LLC Methyl linolenate | 301-00-8 | 99.5% | 292.46 | 5 G

Supplier: Medchemexpress LLC HY212685G
Methyl linolenate is a polyunsaturated fatty acid (PUFA) that exhibits anti-melanogenesis activity with an IC50 of 60 μM. It is utilized for research into the mechanisms and prevention of oxidation/peroxidation of unsaturated fatty acids. In vitro, Methyl linolenate (50, 100 μM, 48 h) significantly down-regulates microphthalmia-associated transcription factor (MITF) and tyrosinase-related proteins in B16F10 cells.
- Polyunsaturated fatty acid (PUFA)
- Exhibits anti-melanogenesis activity
- Supports research on oxidation/peroxidation prevention
- Down-regulates MITF and tyrosinase-related proteins
- Purity of 99.46%
- Molecular formula C19H32O2
- Appearance as a colorless to light yellow liquid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.