Learn More
Medchemexpress LLC Methyl methanesulfonate | 66-27-3 | MFCD00007557 | 110.13 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00470250G
Encompass_Preferred
Methyl methanesulfonate is an alkylating agent that transfers methyl groups and induces DNA damage. It is a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Transfers methyl groups and induces DNA damage
  • Biochemical reagent for life science research
  • Induces cytotoxicity against human MCF7 cells
  • Upregulates phosphorylated p53, RIPK and ROS
  • Induces necroptosis in A549 cells
Catalog No. 50-004-46055