Learn More
Medchemexpress LLC Methyl methanesulfonate (Standard) | 66-27-3 | 110.13 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW004702100G
Encompass_Preferred
Methyl methanesulfonate is an alkylating agent that transfers methyl groups and induces DNA damage. It is a biochemical reagent used as a biological material or organic compound for life science-related research. This product is for research use only and not sold to patients.
  • Transfers methyl groups
  • Induces DNA damage
  • Used as a biochemical reagent
  • Suitable for life science-related research
Catalog No. 50-004-46020