Learn More
Medchemexpress LLC Methyl N-[[(5S)-3-[3-fluoro-4-(2-oxa-6-azaspiro[3.3]heptan-6-yl)phenyl]-2-oxo-1,3-oxazolidin-5-yl] | 2071265-08-0 | 99.7% | 365.36 g/mol | C17H20FN3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1393985MG
Encompass_Preferred
TBI-223 is an orally active oxazolidinone antibiotic research compound investigated for activity against Mycobacterium tuberculosis and other bacterial pathogens. It has been evaluated in preclinical models and shows efficacy in mouse models of bloodstream, skin, and bone infections. The compound is provided for laboratory research in powder and DMSO solution formats.
  • Orally active oxazolidinone antibiotic.
  • Demonstrated activity against Mycobacterium tuberculosis in preclinical studies.
  • Efficacy shown in MRSA mouse models of bloodstream, skin, and bone infection.
  • High purity suitable for research applications (reported ~99.7%).
  • Available as solid powder and as DMSO solution for in vitro and in vivo studies.
  • Molecular formula C17H20FN3O5; molecular weight 365.36 g/mol.
  • Supplied for research use only; not for human therapeutic use.
Catalog No. 50-002-21058