Learn More
Medchemexpress LLC Methyl nadic anhydri 100g
SDP
Supplier:  Medchemexpress LLC HYW106322100G
Encompass_Preferred
Methyl nadic anhydride is an epoxy resin embedding agent used for embedding biological tissue sections
Catalog No. 50-002-04914