Learn More
Medchemexpress LLC Methyl nicotinate | 93-60-7 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB16951G
Encompass_Preferred
Methyl nicotinate (Nicotinic acid methyl ester) is an orally active vasodilator with analgesic activity. When applied topically to the skin, it causes local skin erythema. It is used as an active ingredient in over-the-counter topical preparations for the study of muscle and joint pain.
  • Orally active vasodilator.
  • Analgesic activity.
  • Causes local skin erythema when applied topically.
  • Used in over-the-counter topical preparations for muscle and joint pain.
  • Promoted microvasodilation at 5 mg/mL/egg in a Chorioallantoic Membrane (CAM) model.
  • Significantly reduced writhes induced by acetic acid in mice, showing similar analgesic effect to Aspirin at 10 mg/kg.
  • Significantly prolonged latency of mice's response to thermal pain in a hot plate test.
Catalog No. 50-004-13308