Learn More
Medchemexpress LLC Methyl nonadecanoate | 1731-94-8 | 312.53 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW004262100MG
Encompass_Preferred
Methyl nonadecanoate is an internal standard used to determine the fatty acid methyl ester (FAME) content in biodiesel.
  • Used as an internal standard
  • Determines fatty acid methyl ester (FAME) content in biodiesel
Catalog No. 50-004-47631