Learn More
Medchemexpress LLC Methyl paraben | 99-76-3 | 152.15 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN034950G
Encompass_Preferred
Methyl paraben is a standardized allergen isolated from Yunnan hemlock (Tsuga dumosa). This compound is widely used as a stable, non-volatile preservative. It is known to increase histamine release and regulate cellular immunity, block sodium channels, and prevent ischemia-reperfusion injury.
  • Standardized allergen
  • Stable, non-volatile preservative
  • Increases histamine release and cellular immunity regulation
  • Blocks sodium channels
  • Prevents ischemia-reperfusion injury
  • Reduces basal lipolysis and insulin-stimulated glucose uptake in fat cells
  • Inhibits follicle growth
  • Improves motor deficits
Catalog No. 50-003-89313