Learn More
Medchemexpress LLC Methyl paraben-d4 | 362049-51-2 | 156.17 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0349S5MG
Encompass_Preferred
Methyl paraben-d4 is the deuterium labeled Methyl Paraben. Methyl Paraben, isolated from the barks of Tsuga dumosa, is the methyl ester of p-hydroxybenzoic acid and a standardized chemical allergen. Methyl Paraben is a stable, non-volatile compound used as an antimicrobial preservative in foods, agents, and cosmetics. The physiologic effect of Methyl Paraben involves increased histamine release and cell-mediated immunity.
  • It is a deuterium labeled methyl paraben.
  • It is a stable, non-volatile compound.
  • It can be used as an antimicrobial preservative in foods, agents, and cosmetics.
  • It can be used as a tracer.
  • It can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-90065