Learn More
Medchemexpress LLC Methyl pheophorbide a | 5594-30-9 | 606.71 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN109021MG
Encompass_Preferred
Methyl pheophorbide a is a natural product and one of the chlorophyll-a derivatives. It acts as a photosensitizer that generates singlet oxygen and reactive oxygen species (ROS) when exposed to light of a specific wavelength, which subsequently leads to cell death.
  • Natural product
  • Chlorophyll-a derivative
  • Photosensitizer that produces singlet oxygen and reactive oxygen species (ROS) under light of specific wavelength
  • Leads to cell death
Catalog No. 50-004-15595