Learn More
Medchemexpress LLC Methyl picolinate | 2459-07-6 | 137.14 | 1 ML

Supplier: Medchemexpress LLC HYW00545810MM/1ML
Methyl picolinate is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Purity of 99.91%
- For research use only
- Appears as solid below 18.7°C and liquid above 18.7°C
- Colorless to light yellow appearance
- Soluble in DMSO (≥ 100 mg/mL)
- Store at -80°C for 6 months; -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.