Learn More
TARGETMOL CHEMICALS INC Methyl protodioscin 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T377120MG
Encompass_Preferred
Also available in 1 mL and bulk. Please contact Fisher for quotes. Methyl protodioscin (Smilax saponin B) potentially increase HDL cholesterol while reducing LDL cholesterol and triglycerides. Methyl protodioscin has antitumor property. Methyl protodioscin induced apoptotic process in human A549 cells is closely associated with Mitochondrial membrane potential, caspase-3, and mitochondrial cytochrome c. Purity 99.57%
Catalog No. 50-287-8790