Learn More
Medchemexpress LLC Methyl S---N-Z-aziri 25g | 104597-98-0 | 235.24 g/mol | C12H13NO4 | 25 G
SDP
Supplier:  Medchemexpress LLC HY11702025G
Encompass_Preferred
Methyl (S)-(-)-N-Z-aziridine-2-carboxylate is a chiral aziridine methyl ester used as a synthetic intermediate in pharmaceutical and organic synthesis. It supplies an enantiomerically defined building block for stereoselective transformations and is intended for research and development applications where controlled reactivity of aziridines is required.
  • Chiral (S)-enantiomer suitable for stereoselective synthesis.
  • High listed purity (98.0%) for reliable reactions.
  • Stable under recommended storage: -20°C, sealed and dry.
  • Useful as a precursor for the preparation of amino acid derivatives and other nitrogen-containing scaffolds.
  • Provided as a solid reagent compatible with common organic solvents.
Catalog No. 50-003-07610