Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl S---N-Z-aziri 5g | 104597-98-0 | 5 G
SDP

Supplier:  Medchemexpress LLC HY1170205G

Encompass_Preferred

Methyl (S)-(-)-N-Z-aziridine-2-carboxylate is a chiral aziridine-containing synthetic intermediate used in pharmaceutical and organic synthesis. It is supplied as a liquid (colorless to light yellow) and commonly used as a building block in stereoselective transformations.

  • Chiral aziridine building block for stereoselective synthesis.
  • High purity (98.0%) suitable for synthetic applications.
  • Molecular formula C12H13NO4 and molecular weight 235.24.
  • Available in multiple pack sizes, including 5 g.
  • Liquid appearance, colorless to light yellow; soluble in common organic solvents.

Catalog No. 50-003-08380


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.