Learn More
Medchemexpress LLC Methyl sinapate | 20733-94-2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN3265
Encompass_Preferred
Methyl sinapate (MSA) is a hydroxycinnamic acid and a natural UV screening agent. With a molecular formula of C12H14O5 and a molecular weight of 238.24, it is primarily intended for research use.
  • Solid appearance
  • Pale purple to purple color
  • Soluble in DMSO
  • Initial source: Microorganisms (Fusarium oxysporum)
  • High purity: 95.0%
  • For research use only
Catalog No. 50-004-21923