Learn More
Selleck Chemical LLC Methyl syringate S4787-25mg
SDP
Supplier:  Selleck Chemical LLC S478725MG
Encompass_Preferred
Methyl syringate (Syringic Acid Methyl Ester) a plant phenolic compound is a specific and selective TRPA1 agonist
Catalog No. 50-311-1462